3',5'-Dichlorbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

011481-500MG
FLUO011481-500MGEA 0 EUR
FLUO011481-500MG
3',5'-Dichlorbiphenyl-3-carbonsäure
3',5'-Dichlorbiphenyl-3-carbonsäure
Formel: C₁₃H₈Cl₂O₂
CAS-Nummer: 380228-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR