3',5'-Dichlorbiphenyl-4-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
3',5'-Dichloro-[1,1'-biphenyl]-4-carboxylic acid
011482-500MG
FLUO011482-500MGEA
0
EUR
FLUO011482-500MG
3',5'-Dichlorbiphenyl-4-carbonsäure
3',5'-Dichlorbiphenyl-4-carbonsäure
Formel:
C₁₃H₈Cl₂O₂ Molecular Weight: 267,11 g/mol |
MDL:
MFCD03426496 CAS-Nummer: 190911-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...