4'-Chlorbiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Chloro-[1,1'-biphenyl]-2-carboxylic acid, 4'-Chloro[1,1'-biphenyl]-2-carboxylic acid

011496.500MG 011496-1G
FLUO011496.500MGEA 0 EUR
FLUO011496.500MG FLUO011496-1G
4'-Chlorbiphenyl-2-carbonsäure
4'-Chlorbiphenyl-2-carbonsäure
Formel: C₁₃H₉ClO₂
Molecular Weight: 232,67 g/mol
MDL: MFCD03426504
CAS-Nummer: 7079-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR