4'-Formylbiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

011501-500MG
FLUO011501-500MGEA 0 EUR
FLUO011501-500MG
4'-Formylbiphenyl-2-carbonsäure
4'-Formylbiphenyl-2-carbonsäure
Formel: C₁₄H₁₀O₃
MDL: MFCD03426394
CAS-Nummer: 112804-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR