1-Boc-2-cyanpiperidin

Lieferant: FLUOROCHEM

Synonyme: tert-Butyl 2-cyanopiperidine-1-carboxylate

011512-5G
FLUO011512-5GEA 82.1 EUR
FLUO011512-5G
1-Boc-2-cyanpiperidin
1-Boc-2-cyanpiperidin
Formel: C₁₁H₁₈N₂O₂
Molecular Weight: 210,27 g/mol
MDL: MFCD01861221
CAS-Nummer: 153749-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR