(S)-(-)-1-Boc-3-methylpiperazin

Lieferant: FLUOROCHEM

Synonyme: (S)-tert-Butyl 3-methylpiperazine-1-carboxylate, (3S)-3-Methylpiperazine, N1-BOC protected

011516-5G
FLUO011516-5GEA 0 EUR
FLUO011516-5G
(S)-(-)-1-Boc-3-methylpiperazin
(S)-(-)-1-Boc-3-methylpiperazin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD02683204
CAS-Nummer: 147081-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR