(R)-(+)-1-Boc-3-methylpiperazin

Lieferant: FLUOROCHEM

Synonyme: (3R)-3-Methylpiperazine, N1-BOC protected, (R)-1-Boc-3-Methylpiperazine, (R)-1-Boc-3-methyl-piperazine

011517-5G
FLUO011517-5GEA 79.1 EUR
FLUO011517-5G
(R)-(+)-1-Boc-3-methylpiperazin
(R)-(+)-1-Boc-3-methylpiperazin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD02683205
CAS-Nummer: 163765-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR