1-Boc-3-methylpiperazin

Lieferant: FLUOROCHEM

Synonyme: tert-Butyl 3-methylpiperazine-1-carboxylate, 3-Methylpiperazine, N1-BOC protected

011518-5G
FLUO011518-5GEA 67.1 EUR
FLUO011518-5G
1-Boc-3-methylpiperazin
1-Boc-3-methylpiperazin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
MDL: MFCD03001706
CAS-Nummer: 120737-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR