3-(Boc-amino)-3-(3-chlorphenyl)propionsäure

Lieferant: FLUOROCHEM

Synonyme: 3-tert-Butoxycarbonylamino-3-(3-chlorophenyl)propionic acid, 3-[(tert-Butoxycarbonyl)amino]-3-(3-chlorophenyl)-propanoic acid, 3-tert-butoxycarbonylamino-3-(3-chlorophenyl)propionic acid

011536-10G
FLUO011536-10GEA 330 EUR
FLUO011536-10G
3-(Boc-amino)-3-(3-chlorphenyl)propionsäure
3-(Boc-amino)-3-(3-chlorphenyl)propionsäure
Formel: C₁₄H₁₈ClNO₄
Molecular Weight: 299.76 g/mol
MDL: MFCD02090710
CAS-Nummer: 284493-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR