4-(Boc-amino)piperidin

Lieferant: FLUOROCHEM

Synonyme: 4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine

011547-5G 011547-25G
FLUO011547-5GEA 51.8 EUR
FLUO011547-5G FLUO011547-25G
4-(Boc-amino)piperidin
4-(Boc-amino)piperidin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD00798171
CAS-Nummer: 73874-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR