16-Brom-1-hexadecanol

Lieferant: FLUOROCHEM

Synonyme: 16-Bromohexadecan-1-ol

011575-5G
FLUO011575-5GEA 1670 EUR
FLUO011575-5G
16-Brom-1-hexadecanol
16-Brom-1-hexadecanol
Formel: HO(CH₂)₁₅CH₂Br
Molecular Weight: 321,34 g/mol
MDL: MFCD01076224
CAS-Nummer: 59101-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR