14-Brom-1-tetradecanol
Lieferant: FLUOROCHEM
011577-5G
FLUO011577-5GEA
0
EUR
FLUO011577-5G
14-Brom-1-tetradecanol
14-Brom-1-tetradecanol
Formel:
HO(CH₂)₁₃CH₂Br |
MDL:
MFCD01076225 CAS-Nummer: 72995-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...