5-Brom-2,4-difluoranilin

Lieferant: FLUOROCHEM

011582-5G
FLUO011582-5GEA 31.1 EUR
FLUO011582-5G
5-Brom-2,4-difluoranilin
5-Brom-2,4-difluoranilin
Formel: C₆H₄BrF₂N
Molecular Weight: 208,0000 g/mol
MDL: MFCD03426368
CAS-Nummer: 452-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR