2-Brom-6-formylpyridin

Lieferant: FLUOROCHEM

Synonyme: 6-Bromopyridine-2-carboxaldehyde, 6-Bromopyridine-2-carbaldehyde, 6-Bromo-2-pyridinecarboxaldehyde

011583-5G
FLUO011583-5GEA 55.4 EUR
FLUO011583-5G
2-Brom-6-formylpyridin
2-Brom-6-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
MDL: MFCD02683546
CAS-Nummer: 34160-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR