6-Brompiperonal
Lieferant: FLUOROCHEM
011589-10G
011589-100G
FLUO011589-10GEA
165
EUR
FLUO011589-10G
FLUO011589-100G
6-Brompiperonal
6-Brompiperonal
Formel:
C₈H₅BrO₃ |
MDL:
MFCD00022952 CAS-Nummer: 15930-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...