7-Bromindol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
7-Bromo-1H-indole
011592-1G
FLUO011592-1GEA
45.9
EUR
FLUO011592-1G
7-Bromindol
7-Bromindol
Formel:
C₈H₆BrN Molecular Weight: 196,05 g/mol |
MDL:
MFCD00799492 CAS-Nummer: 51417-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...