α-Brom-3-nitro-p-tolylsäure

Lieferant: FLUOROCHEM

Synonyme: α-Brom-3-nitro-p-toluylsäure, 4-(Brommethyl)-3-nitrobenzoesäure

011593-50G
FLUO011593-50GEA 0 EUR
FLUO011593-50G
α-Brom-3-nitro-p-tolylsäure
α-Brom-3-nitro-p-tolylsäure
Formel: C₈H₆BrNO₄
Molecular Weight: 260,04 g/mol
Schmelzpunkt: 132 °C
MDL: MFCD00216575
CAS-Nummer: 55715-03-2
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR