Di-tert-butyliminodicarboxylat

Lieferant: FLUOROCHEM

011599-50G
FLUO011599-50GEA 0 EUR
FLUO011599-50G
Di-tert-butyliminodicarboxylat
Di-tert-butyliminodicarboxylat
Formel: C₁₀H₁₉NO₄
MDL: MFCD00043309
CAS-Nummer: 51779-32-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR