4-Chlorisatosäureanhydrid

Lieferant: FLUOROCHEM

Synonyme: 7-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione, 7-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione

011636-10G
FLUO011636-10GEA 143 EUR
FLUO011636-10G
4-Chlorisatosäureanhydrid
4-Chlorisatosäureanhydrid
Formel: C₈H₄ClNO₃
Molecular Weight: 197,58 g/mol
MDL: MFCD00090431
CAS-Nummer: 40928-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR