4-(4-Chlorphenyl)-4-piperidinol

Lieferant: FLUOROCHEM

011645-50G
FLUO011645-50GEA 0 EUR
FLUO011645-50G
4-(4-Chlorphenyl)-4-piperidinol
4-(4-Chlorphenyl)-4-piperidinol
Formel: C₁₁H₁₄ClNO
MDL: MFCD00006001
CAS-Nummer: 39512-49-7
EINECS: 254-479-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR