2,6-Dichlor-5-fluornicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 2,6-Dichloro-5-fluoropyridine-3-carboxylic acid

011675-25G
FLUO011675-25GEA 57.9 EUR
FLUO011675-25G
2,6-Dichlor-5-fluornicotinsäure
2,6-Dichlor-5-fluornicotinsäure
Formel: C₆H₂Cl₂FNO₂
Molecular Weight: 209,99 g/mol
MDL: MFCD00799517
CAS-Nummer: 82671-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR