2,7-Dichlorchinoxalin
Lieferant: FLUOROCHEM
011680-5G
FLUO011680-5GEA
2070
EUR
FLUO011680-5G
2,7-Dichlorchinoxalin
2,7-Dichloroquinoxaline
Formel:
C₈H₄Cl₂N₂ Molecular Weight: 199,04 g/mol |
MDL:
MFCD03426386 CAS-Nummer: 59489-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...