2,3-Dimethoxy-p-benzochinon

Lieferant: FLUOROCHEM

011693-1G
FLUO011693-1GEA 1090 EUR
FLUO011693-1G
2,3-Dimethoxy-p-benzochinon
2,3-Dimethoxy-p-benzochinon
Formel: C₈H₈O₄
MDL: MFCD00016366
CAS-Nummer: 3117-02-0
EINECS: 221-489-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR