(1R,2R)-(+)-1,2-Diphenylethylendiamin

Lieferant: FLUOROCHEM

011708-5G 011708-1G
FLUO011708-5GEA 203 EUR
FLUO011708-5G FLUO011708-1G
(1R,2R)-(+)-1,2-Diphenylethylendiamin
(1R,2R)-(+)-1,2-Diphenylethylendiamin
Formel: C₁₄H₁₆N₂
MDL: MFCD00082769
CAS-Nummer: 35132-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR