(1S,2S)-(-)-1,2-Diphenylethylendiamin

Lieferant: FLUOROCHEM

011709-5G
FLUO011709-5GEA 110 EUR
FLUO011709-5G
(1S,2S)-(-)-1,2-Diphenylethylendiamin
(1S,2S)-(-)-1,2-Diphenylethylendiamin
Formel: C₁₄H₁₆N₂
MDL: MFCD00082751
CAS-Nummer: 29841-69-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR