5-Hydroxy-2-nitrobenzaldehyd

Lieferant: FLUOROCHEM
Achtung

011765-50G
FLUO011765-50GEA 0 EUR
FLUO011765-50G
5-Hydroxy-2-nitrobenzaldehyd
5-Hydroxy-2-nitrobenzaldehyd
Formel: C₇H₅NO₄
Molecular Weight: 167,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007332
CAS-Nummer: 42454-06-8
EINECS: 255-832-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR