5-Hydroxy-DL-tryptophan
Lieferant: FLUOROCHEM
011780-10G
FLUO011780-10GEA
0
EUR
FLUO011780-10G
5-Hydroxy-DL-tryptophan
5-Hydroxy-DL-tryptophan
Formel:
C₁₁H₁₂N₂O₃ Schmelzpunkt: 301 °C Dichte: 1,405 g/cm³ (20 °C) |
MDL:
MFCD00005651 CAS-Nummer: 56-69-9 EINECS: 200-284-8 Merck Index: 13,04873 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...