2-Methoxy-4-nitrobenzaldehyd

Lieferant: FLUOROCHEM

011797-5G
FLUO011797-5GEA 446 EUR
FLUO011797-5G
2-Methoxy-4-nitrobenzaldehyd
2-Methoxy-4-nitrobenzaldehyde
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
MDL: MFCD02683052
CAS-Nummer: 136507-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR