3-Methylisatosäureanhydrid
Lieferant: FLUOROCHEM
Synonyme:
8-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione
011816-10G
FLUO011816-10GEA
936
EUR
FLUO011816-10G
3-Methylisatosäureanhydrid
3-Methylisatosäureanhydrid
Formel:
C₉H₇NO₃ Molecular Weight: 177,1568 g/mol |
MDL:
MFCD03426399 CAS-Nummer: 66176-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...