3-Methylglutarsäure

Lieferant: FLUOROCHEM

011821-100G
FLUO011821-100GEA 516 EUR
FLUO011821-100G
3-Methylglutarsäure
3-Methylglutarsäure
Formel: C₆H₁₀O₄
Molecular Weight: 146,1412 g/mol
MDL: MFCD00002727
CAS-Nummer: 626-51-7
EINECS: 210-951-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR