N-Phenylglycin

Lieferant: FLUOROCHEM

Synonyme: 2-Anilinoacetic acid, 2-(Phenylamino)acetic acid

011837-1KG
FLUO011837-1KGEA 0 EUR
FLUO011837-1KG
N-Phenylglycin
N-Phenylglycin
Formel: C₈H₉NO₂
Molecular Weight: 151,17 g/mol
MDL: MFCD00014009
CAS-Nummer: 103-01-5
EINECS: 203-070-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR