Tetrahydropyran-4-essigsäure

Lieferant: FLUOROCHEM

Synonyme: Tetrahydropyranyl-4-acetic acid

011896-1G
FLUO011896-1GEA 48.8 EUR
FLUO011896-1G
Tetrahydropyran-4-essigsäure
Tetrahydropyran-4-essigsäure
Formel: C₇H₁₂O₃
Molecular Weight: 144,1700 g/mol
MDL: MFCD01631204
CAS-Nummer: 85064-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR