3-Formyl-6-methylchromon

Lieferant: CHEMPUR

Synonyme: 6-Methyl-4-oxo-4H-chromene-3-carbaldehyde

FL12002.POR 012002-10G 012002-25G
CHMPFL12002.POREA 0 EUR
CHMPFL12002.POR FLUO012002-10G FLUO012002-25G
3-Formyl-6-methylchromon
3-Formyl-6-methylchromon
Formel: C₁₁H₈O₃
Molecular Weight: 188.1794 g/mol
MDL: MFCD00138943
CAS-Nummer: 42059-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR