6-Fluorchromon

Lieferant: FLUOROCHEM

Synonyme: 6-Fluoro-4H-chromen-4-one

012053-10G
FLUO012053-10GEA 0 EUR
FLUO012053-10G
6-Fluorchromon
6-Fluorchromon
Formel: C₉H₅FO₂
Molecular Weight: 164,14 g/mol
MDL: MFCD03094002
CAS-Nummer: 105300-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR