5-(5-Fluor-2-hydroxyphenyl)isoxazol
Lieferant: FLUOROCHEM
Synonyme:
4-Fluoro-2-(5-isoxazolyl)phenol, 5-(5-Fluoro-2-hydroxyphenyl)isoxazole
012150-5G
FLUO012150-5GEA
142
EUR
FLUO012150-5G
5-(5-Fluor-2-hydroxyphenyl)isoxazol
5-(5-Fluor-2-hydroxyphenyl)isoxazol
Formel:
C₉H₆FNO₂ |
MDL:
MFCD03070563 CAS-Nummer: 288401-62-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...