5-Brom-2-fluor-3-methylpyridin
Lieferant: FLUOROCHEM
Synonyme:
5-Brom-2-fluor-3-picolin
012711-1G
FLUO012711-1GEA
31.1
EUR
FLUO012711-1G
5-Brom-2-fluor-3-methylpyridin
5-Brom-2-fluor-3-methylpyridin
Formel:
C₆H₅BrFN Molecular Weight: 190,01 g/mol |
MDL:
MFCD03095091 CAS-Nummer: 29312-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...