2-Amino-6-chlorpyridin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 6-Chlorpyridin-2-amin, 6-Chlor-2-pyridylamin

012728-5G
FLUO012728-5GEA 47.9 EUR
FLUO012728-5G
2-Amino-6-chlorpyridin
2-Amino-6-chlorpyridin
Formel: C₅H₅ClN₂
Molecular Weight: 128,56 g/mol
MDL: MFCD00234068
CAS-Nummer: 45644-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR