2-Brom-4-chlorpyridin

Lieferant: FLUOROCHEM

012729 012729-1G 012729-5G
FLUO012729EA 0 EUR
FLUO012729 FLUO012729-1G FLUO012729-5G
2-Brom-4-chlorpyridin
2-Bromo-4-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
MDL: MFCD03840757
CAS-Nummer: 22918-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR