2-Brom-5-chlorpyridin

Lieferant: FLUOROCHEM

012730-100G 012730-25G
FLUO012730-100GEA 200 EUR
FLUO012730-100G FLUO012730-25G
2-Brom-5-chlorpyridin
2-Bromo-5-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
MDL: MFCD00234006
CAS-Nummer: 40473-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR