4-Hydroxy-3-nitropyridin
Lieferant: FLUOROCHEM
Synonyme:
3-Nitro-4-pyridinol
012733-25G
FLUO012733-25GEA
39.6
EUR
FLUO012733-25G
4-Hydroxy-3-nitropyridin
4-Hydroxy-3-nitropyridine
Formel:
C₅H₄N₂O₃ Molecular Weight: 140,10 g/mol |
MDL:
MFCD00128874 CAS-Nummer: 5435-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...