2,6-Dichlor-5-fluornicotinonitril

Lieferant: FLUOROCHEM

Synonyme: 2,6-Dichloro-3-cyano-5-fluoropyridine, 2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile

012735-25G
FLUO012735-25GEA 52.5 EUR
FLUO012735-25G
2,6-Dichlor-5-fluornicotinonitril
2,6-Dichlor-5-fluornicotinonitril
Formel: C₆HCl₂FN₂
Molecular Weight: 190,99 g/mol
MDL: MFCD01863289
CAS-Nummer: 82671-02-1
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR