2,6-Dichlor-3-nitropyridin

Lieferant: FLUOROCHEM

012736-25G
FLUO012736-25GEA 0 EUR
FLUO012736-25G
2,6-Dichlor-3-nitropyridin
2,6-Dichloro-3-nitropyridine
Formel: C₅H₂Cl₂N₂O₂
Molecular Weight: 192,99 g/mol
Schmelzpunkt: 67…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006234
CAS-Nummer: 16013-85-7
EINECS: 240-151-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR