4-Chlor-2-fluorbenzaldehyd

Lieferant: FLUOROCHEM
Achtung

013011-5G 013011-25G
FLUO013011-5GEA 0 EUR
FLUO013011-5G FLUO013011-25G
4-Chlor-2-fluorbenzaldehyd
4-Chloro-2-fluorobenzaldehyde
Formel: C₇H₄ClFO
Molecular Weight: 158.56 g/mol
MDL: MFCD00143282
CAS-Nummer: 61072-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR