4-Chlor-2-fluorbenzaldehyd
Lieferant: FLUOROCHEM
|
Achtung
|
013011-5G
013011-25G
FLUO013011-5GEA
0
EUR
FLUO013011-5G
FLUO013011-25G
4-Chlor-2-fluorbenzaldehyd
4-Chloro-2-fluorobenzaldehyde
Formel:
C₇H₄ClFO Molecular Weight: 158.56 g/mol |
MDL:
MFCD00143282 CAS-Nummer: 61072-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...