6-Fluor-o-anisaldehyd
Lieferant: FLUOROCHEM
Synonyme:
6-Fluor-2-anisaldehyd, 2-Fluor-6-methoxybenzaldehyd
013017-1G
013017-5G
FLUO013017-1GEA
31.1
EUR
FLUO013017-1G
FLUO013017-5G
6-Fluor-o-anisaldehyd
6-Fluor-o-anisaldehyd
Formel:
C₈H₇FO₂ Molecular Weight: 154.14 g/mol |
MDL:
MFCD01090998 CAS-Nummer: 146137-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...