3,5-Dichlorbenzylalkohol
Lieferant: FLUOROCHEM
013039-5G
013039-25G
FLUO013039-5GEA
39.9
EUR
FLUO013039-5G
FLUO013039-25G
3,5-Dichlorbenzylalkohol
3,5-Dichlorbenzylalkohol
Formel:
C₇H₆Cl₂O Siedepunkt: 264 °C (1013 hPa) Schmelzpunkt: 82 °C |
MDL:
MFCD00004634 CAS-Nummer: 60211-57-6 EINECS: 262-109-1 UN: 2250 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...