2-(Trifluormethoxy)phenylboronsäure
Lieferant: FLUOROCHEM
013043-1G
013043-5G
FLUO013043-1GEA
31.1
EUR
FLUO013043-1G
FLUO013043-5G
2-(Trifluormethoxy)phenylboronsäure
2-(Trifluormethoxy)phenylboronsäure
Formel:
C₇H₆BF₃O₃ |
MDL:
MFCD01320763 CAS-Nummer: 175676-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...