2-(Trifluormethoxy)phenylboronsäure

Lieferant: FLUOROCHEM

013043-1G 013043-5G
FLUO013043-1GEA 31.1 EUR
FLUO013043-1G FLUO013043-5G
2-(Trifluormethoxy)phenylboronsäure
2-(Trifluormethoxy)phenylboronsäure
Formel: C₇H₆BF₃O₃
MDL: MFCD01320763
CAS-Nummer: 175676-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR