3-(Trifluormethoxy)thiophenol

Lieferant: FLUOROCHEM

Synonyme: 3-(Trifluormethoxy)phenylmercaptan, 3-(Trifluormethoxy)benzolthiol

013053-1G 013053-5G
FLUO013053-1GEA 136 EUR
FLUO013053-1G FLUO013053-5G
3-(Trifluormethoxy)thiophenol
3-(Trifluormethoxy)thiophenol
Formel: C₇H₅F₃OS
Molecular Weight: 194.18 g/mol
MDL: MFCD01320799
CAS-Nummer: 220239-66-7
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR