2-Brom-5-(trifluormethyl)benzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Brom-5-(trifluormethyl)benzol-1-sulfonylchlorid, 2-Brom-5-(trifluormethyl)benzolsulfochlorid, 2-Brom-5-(trifluormethyl)benzolsulfonsäurechlorid

013084-5G 013084-25G
FLUO013084-5GEA 249 EUR
FLUO013084-5G FLUO013084-25G
2-Brom-5-(trifluormethyl)benzolsulfonylchlorid
2-Brom-5-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃BrClF₃O₂S
Siedepunkt: 230…231 °C (1013 hPa)
MDL: MFCD03094395
CAS-Nummer: 176225-08-4
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR